Assay Details
| Gene Symbol: eef2.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203924.1 | NP_989255.1 | 5 - 6 | 917 | 83 | |
| GenBank mRNA | BC063919.1 | - | 5 - 6 | 917 | 83 | |
| BC084061.1 | - | 5 - 6 | 857 | 83 | ||
| CR848340.2 | - | 4 - 5 | 791 | 83 | ||
More Information
Set Membership: |
|

eef2.1
Genomic Map