Assay Details
| Gene Symbol: slc49a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203929.1 | NP_989260.1 | 1 - 2 | 319 | 82 | |
| XM_012958339.2 | XP_012813793.1 | 1 - 2 | 618 | 82 | ||
| GenBank mRNA | BC063895.1 | - | 1 - 2 | 319 | 82 | |
More Information
Set Membership: |
|

slc49a3
Genomic Map