Assay Details
| Gene Symbol: slc49a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203929.1 | NP_989260.1 | 7 - 8 | 1186 | 67 | |
| XM_012958339.2 | XP_012813793.1 | 7 - 8 | 1485 | 67 | ||
| GenBank mRNA | BC063895.1 | - | 7 - 8 | 1186 | 67 | |
| CR761997.2 | - | 7 - 8 | 1184 | 67 | ||
More Information
Set Membership: |
|

slc49a3
Genomic Map