Assay Details
| Gene Symbol: tmed7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203930.1 | NP_989261.1 | 2 - 3 | 311 | 86 | |
| XM_012958340.2 | XP_012813794.1 | 2 - 3 | 503 | 86 | ||
| XM_012958341.2 | XP_012813795.1 | 2 - 3 | 262 | 86 | ||
| GenBank mRNA | BC064279.1 | - | 2 - 3 | 311 | 86 | |
| CT025507.2 | - | 1 - 2 | 221 | 86 | ||
More Information
Set Membership: |
|

tmed7
Genomic Map