Assay Details
Gene Symbol: eef1g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203931.1 | NP_989262.1 | 6 - 7 | 657 | 60 | |
XM_012953583.1 | XP_012809037.1 | 5 - 6 | 712 | 60 | ||
GenBank mRNA | BC064264.1 | - | 6 - 7 | 657 | 60 | |
BC080966.1 | - | 4 - 5 | 517 | 60 | ||
BC154938.1 | - | 4 - 5 | 455 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
