Assay Details
| Gene Symbol: lrr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203947.1 | NP_989278.1 | 2 - 3 | 356 | 81 | |
| GenBank mRNA | BC064158.1 | - | 2 - 3 | 356 | 81 | |
| CR761436.2 | - | 2 - 3 | 305 | 81 | ||
| CU024931.1 | - | 2 - 3 | 312 | 81 | ||
More Information
Set Membership: |
|

lrr1
Genomic Map