Assay Details
Gene Symbol: adap1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203955.1 | NP_989286.1 | 10 - 11 | 1221 | 72 | |
XM_012960771.2 | XP_012816225.1 | 10 - 11 | 1468 | 72 | ||
XM_012960772.2 | XP_012816226.1 | 9 - 10 | 1219 | 72 | ||
XM_018092976.1 | XP_017948465.1 | 9 - 10 | 1266 | 72 | ||
XM_018092977.1 | XP_017948466.1 | 9 - 10 | 1205 | 72 | ||
XM_018092979.1 | XP_017948468.1 | 9 - 10 | 2342 | 72 | ||
GenBank mRNA | BC064211.1 | - | 10 - 11 | 1221 | 72 | |
CR760308.2 | - | 10 - 11 | 1142 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
