Assay Details
| Gene Symbol: timm10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203963.2 | NP_989294.1 | 2 - 3 | 147 | 97 | |
| XM_012958374.2 | XP_012813828.1 | 2 - 3 | 242 | 97 | ||
| GenBank mRNA | CR848606.2 | - | 2 - 3 | 147 | 97 | |
More Information
Set Membership: |
|

timm10
Genomic Map