Assay Details
| Gene Symbol: efhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203969.1 | NP_989300.1 | 2 - 3 | 474 | 94 | |
| GenBank mRNA | BC063917.1 | - | 2 - 3 | 474 | 94 | |
| BC076710.1 | - | 2 - 3 | 508 | 94 | ||
| CT030284.1 | - | 2 - 3 | 450 | 94 | ||
More Information
Set Membership: |
|

efhd1
Genomic Map