Assay Details
| Gene Symbol: slc34a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203971.3 | NP_989302.2 | 9 - 10 | 1080 | 63 | |
| XM_018089588.1 | XP_017945077.1 | 9 - 10 | 1286 | 63 | ||
More Information
Set Membership: |
|

slc34a2
Genomic Map