Assay Details
| Gene Symbol: dhrs13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203980.1 | NP_989311.1 | 2 - 3 | 270 | 99 | |
| GenBank mRNA | BC063926.1 | - | 2 - 3 | 270 | 99 | |
| CR926297.2 | - | 2 - 3 | 322 | 99 | ||
More Information
Set Membership: |
|

dhrs13
Genomic Map