Assay Details
| Gene Symbol: ppat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203982.1 | NP_989313.1 | 5 - 6 | 732 | 61 | |
| GenBank mRNA | BC064240.1 | - | 5 - 6 | 732 | 61 | |
| CR926382.1 | - | 5 - 6 | 697 | 61 | ||
More Information
Set Membership: |
|

ppat
Genomic Map