Assay Details
| Gene Symbol: rab6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203984.1 | NP_989315.1 | 506 | 66 | ||
| GenBank mRNA | BC064230.1 | - | 506 | 66 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

rab6a| Gene Symbol: rab6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203984.1 | NP_989315.1 | 506 | 66 | ||
| GenBank mRNA | BC064230.1 | - | 506 | 66 | ||
Set Membership: |
|