Assay Details
| Gene Symbol: szrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203987.1 | NP_989318.1 | 364 | 67 | ||
| GenBank mRNA | BC064214.1 | - | 364 | 67 | ||
| CT030456.1 | - | 1 - 2 | 370 | 67 | ||
More Information
Set Membership: |
|

szrd1
Genomic Map