Assay Details
| Gene Symbol: LOC394967 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204010.1 | NP_989341.1 | 10 - 11 | 1189 | 60 | |
| XM_012966309.1 | XP_012821763.1 | 11 - 12 | 1306 | 60 | ||
| GenBank mRNA | BC064243.1 | - | 10 - 11 | 1189 | 60 | |
| BC080957.1 | - | 10 - 11 | 1178 | 60 | ||
| CR760232.2 | - | 10 - 11 | 1180 | 60 | ||
More Information
Set Membership: |
|

LOC394967
Genomic Map