Assay Details
| Gene Symbol: LOC394967 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204010.1 | NP_989341.1 | 3 - 4 | 341 | 88 | |
| XM_012966309.1 | XP_012821763.1 | 4 - 5 | 458 | 88 | ||
| GenBank mRNA | BC064243.1 | - | 3 - 4 | 341 | 88 | |
| BC080957.1 | - | 3 - 4 | 330 | 88 | ||
| CR760232.2 | - | 3 - 4 | 332 | 88 | ||
More Information
Set Membership: |
|

LOC394967
Genomic Map