Assay Details
| Gene Symbol: stoml3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204013.1 | NP_989344.1 | 4 - 5 | 271 | 57 | |
| GenBank mRNA | BC064171.1 | - | 4 - 5 | 271 | 57 | |
| CR942640.2 | - | 4 - 5 | 274 | 57 | ||
More Information
Set Membership: |
|

stoml3
Genomic Map