Assay Details
| Gene Symbol: sparc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204016.2 | NP_989347.1 | 6 - 7 | 489 | 75 | |
| GenBank mRNA | AY575077.1 | - | 6 - 7 | 484 | 75 | |
| BC064259.1 | - | 6 - 7 | 484 | 75 | ||
| BC079950.1 | - | 6 - 7 | 489 | 75 | ||
| CT010536.2 | - | 6 - 7 | 484 | 75 | ||
More Information
Set Membership: |
|

sparc
Genomic Map