Assay Details
| Gene Symbol: sdhd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204026.1 | NP_989357.1 | 2 - 3 | 195 | 101 | |
| GenBank mRNA | BC064178.1 | - | 2 - 3 | 195 | 101 | |
| CR848414.2 | - | 2 - 3 | 161 | 101 | ||
More Information
Set Membership: |
|

sdhd
Genomic Map