Assay Details
| Gene Symbol: eif3h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204028.1 | NP_989359.1 | 2 - 3 | 256 | 62 | |
| GenBank mRNA | BC064151.1 | - | 2 - 3 | 256 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

eif3h| Gene Symbol: eif3h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204028.1 | NP_989359.1 | 2 - 3 | 256 | 62 | |
| GenBank mRNA | BC064151.1 | - | 2 - 3 | 256 | 62 | |
Set Membership: |
|