Assay Details
| Gene Symbol: lsm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204040.1 | NP_989371.1 | 3 - 4 | 170 | 86 | |
| XM_012958531.2 | XP_012813985.1 | 3 - 4 | 218 | 86 | ||
| GenBank mRNA | BC064199.1 | - | 3 - 4 | 170 | 86 | |
| BC082491.1 | - | 3 - 4 | 168 | 86 | ||
| CT025508.2 | - | 3 - 4 | 176 | 86 | ||
More Information
Set Membership: |
|

lsm4
Genomic Map