Assay Details
| Gene Symbol: znf207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204045.1 | NP_989376.1 | 1 - 2 | 208 | 90 | |
| GenBank mRNA | BC064165.1 | - | 1 - 2 | 208 | 90 | |
| CR762039.2 | - | 1 - 2 | 194 | 90 | ||
More Information
Set Membership: |
|

znf207
Genomic Map