Assay Details
| Gene Symbol: med11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204048.1 | NP_989379.1 | 2 - 3 | 178 | 80 | |
| GenBank mRNA | BC064722.1 | - | 2 - 3 | 178 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

med11| Gene Symbol: med11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204048.1 | NP_989379.1 | 2 - 3 | 178 | 80 | |
| GenBank mRNA | BC064722.1 | - | 2 - 3 | 178 | 80 | |
Set Membership: |
|