Assay Details
| Gene Symbol: rcn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204058.1 | NP_989389.1 | 4 - 5 | 634 | 80 | |
| XM_012956681.2 | XP_012812135.1 | 4 - 5 | 709 | 80 | ||
| GenBank mRNA | BC064883.1 | - | 4 - 5 | 634 | 80 | |
| CT030617.1 | - | 4 - 5 | 633 | 80 | ||
More Information
Set Membership: |
|

rcn3
Genomic Map