Assay Details
| Gene Symbol: slc25a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204060.3 | NP_989391.1 | 1 - 2 | 291 | 90 | |
| GenBank mRNA | BC064874.1 | - | 1 - 2 | 263 | 90 | |
| CT030484.1 | - | 1 - 2 | 179 | 90 | ||
More Information
Set Membership: |
|

slc25a1
Genomic Map