Assay Details
| Gene Symbol: kbtbd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204065.1 | NP_989396.1 | 2 - 3 | 768 | 87 | |
| GenBank mRNA | BC064867.1 | - | 2 - 3 | 768 | 87 | |
More Information
Set Membership: |
|

kbtbd4
Genomic Map