Assay Details
| Gene Symbol: anp32c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204073.1 | NP_989404.1 | 1 - 2 | 84 | 77 | |
| XM_018091892.1 | XP_017947381.1 | 1 - 2 | 144 | 77 | ||
| GenBank mRNA | BC064864.1 | - | 1 - 2 | 84 | 77 | |
| CR855787.2 | - | 1 - 2 | 79 | 77 | ||
More Information
Set Membership: |
|

anp32c
Genomic Map