Assay Details
| Gene Symbol: atp1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204076.1 | NP_989407.1 | 10 - 11 | 1502 | 77 | |
| GenBank mRNA | BC064884.1 | - | 10 - 11 | 1502 | 77 | |
| BC080463.1 | - | 10 - 11 | 1520 | 77 | ||
| BC161148.1 | - | 10 - 11 | 1502 | 77 | ||
| CR926442.2 | - | 10 - 11 | 1496 | 77 | ||
More Information
Set Membership: |
|

atp1a1
Genomic Map