Assay Details
| Gene Symbol: gata6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204091.1 | NP_989422.1 | 3 - 4 | 1162 | 62 | |
| XM_012964475.2 | XP_012819929.1 | 3 - 4 | 1190 | 62 | ||
| XM_012964476.2 | XP_012819930.1 | 3 - 4 | 1165 | 62 | ||
| GenBank mRNA | AY260735.1 | - | 3 - 4 | 1162 | 62 | |
| CT030595.1 | - | 3 - 4 | 1162 | 62 | ||
More Information
Set Membership: |
|

gata6
Genomic Map