Assay Details
| Gene Symbol: h1f0 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213671.1 | NP_998836.1 | 1 | 949 | 86 | |
| GenBank mRNA | BC067985.1 | - | 1 | 949 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

h1f0| Gene Symbol: h1f0 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213671.1 | NP_998836.1 | 1 | 949 | 86 | |
| GenBank mRNA | BC067985.1 | - | 1 | 949 | 86 | |
Set Membership: |
|