Assay Details
| Gene Symbol: ccno | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213672.1 | NP_998837.1 | 2 - 3 | 604 | 72 | |
| GenBank mRNA | BC067943.1 | - | 2 - 3 | 604 | 72 | |
| CR761847.2 | - | 2 - 3 | 599 | 72 | ||
More Information
Set Membership: |
|

ccno
Genomic Map