Assay Details
| Gene Symbol: atad3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213684.1 | NP_998849.1 | 8 - 9 | 982 | 71 | |
| GenBank mRNA | BC067935.1 | - | 8 - 9 | 982 | 71 | |
More Information
Set Membership: |
|

atad3a
Genomic Map