Assay Details
| Gene Symbol: ag1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213699.1 | NP_998864.1 | 5 - 6 | 442 | 70 | |
| GenBank mRNA | BC067924.1 | - | 5 - 6 | 442 | 70 | |
| CT030434.1 | - | 5 - 6 | 404 | 70 | ||
More Information
Set Membership: |
|

ag1
Genomic Map