Assay Details
| Gene Symbol: chmp2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213701.1 | NP_998866.1 | 4 - 5 | 468 | 111 | |
| GenBank mRNA | BC067988.1 | - | 4 - 5 | 468 | 111 | |
More Information
Set Membership: |
|

chmp2b
Genomic Map