Assay Details
| Gene Symbol: sox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213704.3 | NP_998869.1 | 1 | 1025 | 86 | |
| GenBank mRNA | BC067975.2 | - | 1 | 950 | 86 | |
| CR760314.2 | - | 1 | 773 | 86 | ||
More Information
Set Membership: |
|

sox2
Genomic Map