Assay Details
| Gene Symbol: mcm7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213712.1 | NP_998877.1 | 3 - 4 | 306 | 83 | |
| GenBank mRNA | BC067307.1 | - | 3 - 4 | 306 | 83 | |
| CR855766.2 | - | 3 - 4 | 285 | 83 | ||
More Information
Set Membership: |
|

mcm7
Genomic Map