Assay Details
| Gene Symbol: gtf2a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005107.2 | NP_001005107.1 | 2 - 3 | 198 | 73 | |
| XM_012970312.2 | XP_012825766.1 | 2 - 3 | 248 | 73 | ||
| GenBank mRNA | BC077041.1 | - | 2 - 3 | 199 | 73 | |
More Information
Set Membership: |
|

gtf2a2
Genomic Map