Assay Details
| Gene Symbol: gtf2a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005107.2 | NP_001005107.1 | 4 - 5 | 427 | 92 | |
| XM_012970312.2 | XP_012825766.1 | 4 - 5 | 477 | 92 | ||
| GenBank mRNA | BC077041.1 | - | 4 - 5 | 428 | 92 | |
More Information
Set Membership: |
|

gtf2a2
Genomic Map