Assay Details
| Gene Symbol: fkbp14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005108.1 | NP_001005108.1 | 3 - 4 | 614 | 95 | |
| GenBank mRNA | BC077042.1 | - | 3 - 4 | 614 | 95 | |
| CR761056.2 | - | 3 - 4 | 562 | 95 | ||
More Information
Set Membership: |
|

fkbp14
Genomic Map