Assay Details
| Gene Symbol: sumo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005111.1 | NP_001005111.1 | 3 - 4 | 271 | 96 | |
| GenBank mRNA | BC077048.1 | - | 3 - 4 | 271 | 96 | |
More Information
Set Membership: |
|

sumo1
Genomic Map