Assay Details
| Gene Symbol: nme3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005115.2 | NP_001005115.1 | 3 - 4 | 330 | 89 | |
| XM_012970313.2 | XP_012825767.1 | 3 - 4 | 945 | 89 | ||
| GenBank mRNA | BC077052.1 | - | 3 - 4 | 331 | 89 | |
| CR760174.2 | - | 3 - 4 | 283 | 89 | ||
More Information
Set Membership: |
|

nme3
Genomic Map