Assay Details
| Gene Symbol: rpl31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005118.2 | NP_001005118.1 | 3 - 4 | 253 | 96 | |
| XM_012956773.2 | XP_012812227.1 | 3 - 4 | 287 | 96 | ||
| XM_018091176.1 | XP_017946665.1 | 3 - 4 | 248 | 96 | ||
| GenBank mRNA | BC077057.1 | - | 3 - 4 | 254 | 96 | |
More Information
Set Membership: |
|

rpl31
Genomic Map