Assay Details
| Gene Symbol: ndufab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005125.1 | NP_001005125.1 | 4 - 5 | 516 | 66 | |
| GenBank mRNA | BC077661.1 | - | 4 - 5 | 516 | 66 | |
| CR760725.2 | - | 4 - 5 | 494 | 66 | ||
More Information
Set Membership: |
|

ndufab1
Genomic Map