Assay Details
| Gene Symbol: rpl37a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005137.1 | NP_001005137.1 | 3 - 4 | 233 | 86 | |
| GenBank mRNA | BC077677.1 | - | 3 - 4 | 233 | 86 | |
More Information
Set Membership: |
|

rpl37a
Genomic Map