Assay Details
| Gene Symbol: slc25a22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005430.1 | NP_001005430.1 | 9 - 10 | 977 | 67 | |
| XM_012958836.2 | XP_012814290.1 | 6 - 7 | 3798 | 67 | ||
| GenBank mRNA | BC074507.1 | - | 9 - 10 | 977 | 67 | |
More Information
Set Membership: |
|

slc25a22
Genomic Map