Assay Details
| Gene Symbol: echs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005437.1 | NP_001005437.1 | 5 - 6 | 643 | 70 | |
| XM_018095333.1 | XP_017950822.1 | 5 - 6 | 667 | 70 | ||
| GenBank mRNA | BC074535.1 | - | 5 - 6 | 643 | 70 | |
More Information
Set Membership: |
|

echs1
Genomic Map