Assay Details
| Gene Symbol: odc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005441.1 | NP_001005441.1 | 8 - 9 | 1057 | 64 | |
| GenBank mRNA | BC074547.1 | - | 8 - 9 | 1057 | 64 | |
| CT025275.2 | - | 6 - 7 | 752 | 64 | ||
More Information
Set Membership: |
|

odc1
Genomic Map