Assay Details
| Gene Symbol: cdk9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005448.1 | NP_001005448.1 | 6 - 7 | 1072 | 95 | |
| XM_012968192.2 | XP_012823646.1 | 6 - 7 | 1128 | 95 | ||
| XM_012968193.2 | XP_012823647.1 | 6 - 7 | 1098 | 95 | ||
| GenBank mRNA | BC074560.1 | - | 6 - 7 | 1072 | 95 | |
| CR762201.2 | - | 6 - 7 | 912 | 95 | ||
More Information
Set Membership: |
|

cdk9
Genomic Map