Assay Details
| Gene Symbol: elovl7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005456.1 | NP_001005456.1 | 7 - 8 | 692 | 79 | |
| XM_012958973.1 | XP_012814427.1 | 7 - 8 | 743 | 79 | ||
| GenBank mRNA | BC074576.1 | - | 7 - 8 | 692 | 79 | |
More Information
Set Membership: |
|

elovl7
Genomic Map