Assay Details
| Gene Symbol: abce1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005628.1 | NP_001005628.1 | 10 - 11 | 1093 | 63 | |
| GenBank mRNA | BC074613.1 | - | 10 - 11 | 1093 | 63 | |
| CR760327.2 | - | 10 - 11 | 1048 | 63 | ||
More Information
Set Membership: |
|

abce1
Genomic Map